Butyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450453-25G
APOSPC450453-25GEA 1540 EUR
APOSPC450453-25G
Butyl 1H,1H,2H,2H-perfluorohexyl carbonate
Butyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₁₁H₁₃F₉O₃
Molecular Weight: 364,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334427
CAS-Nummer: 1980086-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR