Butyl 1H,1H,7H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450454-25G
APOSPC450454-25GEA 1540 EUR
APOSPC450454-25G
Butyl 1H,1H,7H-perfluorohexyl carbonate
Butyl 1H,1H,7H-perfluorohexyl carbonate
Formel: C₁₂H₁₂F₁₂O₃
Molecular Weight: 432,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334428
CAS-Nummer: 1980034-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR