Butyl 1H,1H,9H-perfluorononyl carbonate

Lieferant: Apollo Scientific

PC450455-25G
APOSPC450455-25GEA 1540 EUR
APOSPC450455-25G
Butyl 1H,1H,9H-perfluorononyl carbonate
Butyl 1H,1H,9H-perfluorononyl carbonate
Formel: C₁₄H₁₂F₁₆O₃
Molecular Weight: 532,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334429
CAS-Nummer: 1980034-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR