4-Bromo-3,3,4,4-tetrafluorobutyl butyl carbonate

Lieferant: Apollo Scientific

PC450456-25G
APOSPC450456-25GEA 1540 EUR
APOSPC450456-25G
4-Bromo-3,3,4,4-tetrafluorobutyl butyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl butyl carbonate
Formel: C₉H₁₃BrF₄O₃
Molecular Weight: 325,095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334473
CAS-Nummer: 1923065-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR