Butyl hexafluoroisopropyl carbonate

Lieferant: Apollo Scientific

PC450457-25G
APOSPC450457-25GEA 1540 EUR
APOSPC450457-25G
Butyl hexafluoroisopropyl carbonate
Butyl hexafluoroisopropyl carbonate
Formel: C₈H₁₀F₆O₃
Molecular Weight: 268,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334430
CAS-Nummer: 1923267-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR