tert-Butyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450458-25G
APOSPC450458-25GEA 1540 EUR
APOSPC450458-25G
tert-Butyl 2,2,2-trifluoroethyl carbonate
tert-Butyl 2,2,2-trifluoroethyl carbonate
Formel: C₇H₁₁F₃O₃
Molecular Weight: 200,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334431
CAS-Nummer: 305856-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR