tert-Butyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450460-25G
APOSPC450460-25GEA 1540 EUR
APOSPC450460-25G
tert-Butyl 2,2,3,3-tetrafluoropropyl carbonate
tert-Butyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₈H₁₂F₄O₃
Molecular Weight: 232,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334433
CAS-Nummer: 1980035-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR