tert-Butyl 3-fluoropropyl carbonate
Lieferant: Apollo Scientific
PC450461-25G
APOSPC450461-25GEA
1540
EUR
APOSPC450461-25G
tert-Butyl 3-fluoropropyl carbonate
tert-Butyl 3-fluoropropyl carbonate
Formel:
C₈H₁₅FO₃ Molecular Weight: 178,201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334434 CAS-Nummer: 1980034-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...