tert-Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450462-25G
APOSPC450462-25GEA 1540 EUR
APOSPC450462-25G
tert-Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
tert-Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Formel: C₉H₁₁F₇O₃
Molecular Weight: 300,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334435
CAS-Nummer: 1980086-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR