tert-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450463-25G
APOSPC450463-25GEA 1540 EUR
APOSPC450463-25G
tert-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate
tert-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formel: C₉H₁₂F₆O₃
Molecular Weight: 282,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334436
CAS-Nummer: 1980039-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR