tert-Butyl 4,4,5,5,5-pentafluoropentyl carbonate
Lieferant: Apollo Scientific
PC450464-25G
APOSPC450464-25GEA
1540
EUR
APOSPC450464-25G
tert-Butyl 4,4,5,5,5-pentafluoropentyl carbonate
tert-Butyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel:
C₁₀H₁₅F₅O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334437 CAS-Nummer: 1980038-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...