tert-Butyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450465-25G
APOSPC450465-25GEA 1540 EUR
APOSPC450465-25G
tert-Butyl 1H,1H-perfluorohexyl carbonate
tert-Butyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₁H₁₁F₁₁O₃
Molecular Weight: 400,186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334438
CAS-Nummer: 1980086-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR