tert-Butyl 1H,1H,2H,2H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450466-25G
APOSPC450466-25GEA
1540
EUR
APOSPC450466-25G
tert-Butyl 1H,1H,2H,2H-perfluorohexyl carbonate
tert-Butyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel:
C₁₁H₁₃F₉O₃ Molecular Weight: 364,205 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334439 CAS-Nummer: 1980085-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...