tert-Butyl 1H,1H,7H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450467-25G
APOSPC450467-25GEA 1540 EUR
APOSPC450467-25G
tert-Butyl 1H,1H,7H-perfluorohexyl carbonate
tert-Butyl 1H,1H,7H-perfluorohexyl carbonate
Formel: C₁₂H₁₂F₁₂O₃
Molecular Weight: 432,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334440
CAS-Nummer: 1980038-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR