4-Bromo-3,3,4,4-tetrafluorobutyl tert-butyl carbonate

Lieferant: Apollo Scientific

PC450469-25G
APOSPC450469-25GEA 1540 EUR
APOSPC450469-25G
4-Bromo-3,3,4,4-tetrafluorobutyl tert-butyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl tert-butyl carbonate
Formel: C₉H₁₃BrF₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334474
CAS-Nummer: 1980085-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR