tert-Butyl hexafluoroisopropyl carbonate

Lieferant: Apollo Scientific

PC450470-25G
APOSPC450470-25GEA 1540 EUR
APOSPC450470-25G
tert-Butyl hexafluoroisopropyl carbonate
tert-Butyl hexafluoroisopropyl carbonate
Formel: C₈H₁₀F₆O₃
Molecular Weight: 268,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334442
CAS-Nummer: 1980034-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR