sec-Butyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450471-25G
APOSPC450471-25GEA 1540 EUR
APOSPC450471-25G
sec-Butyl 2,2,2-trifluoroethyl carbonate
sec-Butyl 2,2,2-trifluoroethyl carbonate
Formel: C₇H₁₁F₃O₃
Molecular Weight: 200,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334443
CAS-Nummer: 1980085-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR