sec-Butyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450472-25G
APOSPC450472-25GEA 1540 EUR
APOSPC450472-25G
sec-Butyl 2,2,3,3,3-pentafluoropropyl carbonate
sec-Butyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₈H₁₁F₅O₃
Molecular Weight: 250,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334444
CAS-Nummer: 1923065-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR