sec-Butyl 2,2,3,3-tetrafluoropropyl carbonate
Lieferant: Apollo Scientific
PC450473-25G
APOSPC450473-25GEA
1540
EUR
APOSPC450473-25G
sec-Butyl 2,2,3,3-tetrafluoropropyl carbonate
sec-Butyl 2,2,3,3-tetrafluoropropyl carbonate
Formel:
C₈H₁₂F₄O₃ Molecular Weight: 232,173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334445 CAS-Nummer: 1980034-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...