sec-Butyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450478-25G
APOSPC450478-25GEA 1540 EUR
APOSPC450478-25G
sec-Butyl 1H,1H-perfluorohexyl carbonate
sec-Butyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₁H₁₁F₁₁O₃
Molecular Weight: 400,186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334450
CAS-Nummer: 1980085-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR