sec-Butyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450479-25G
APOSPC450479-25GEA 1540 EUR
APOSPC450479-25G
sec-Butyl 1H,1H,2H,2H-perfluorohexyl carbonate
sec-Butyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₁₁H₁₃F₉O₃
Molecular Weight: 364,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334451
CAS-Nummer: 1980034-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR