sec-Butyl 1H,1H,7H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450480-25G
APOSPC450480-25GEA
1540
EUR
APOSPC450480-25G
sec-Butyl 1H,1H,7H-perfluorohexyl carbonate
sec-Butyl 1H,1H,7H-perfluorohexyl carbonate
Formel:
C₁₂H₁₂F₁₂O₃ Molecular Weight: 432,203 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334452 CAS-Nummer: 1980034-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...