sec-Butyl 1H,1H,9H-perfluorononyl carbonate
Lieferant: Apollo Scientific
PC450481-25G
APOSPC450481-25GEA
1540
EUR
APOSPC450481-25G
sec-Butyl 1H,1H,9H-perfluorononyl carbonate
sec-Butyl 1H,1H,9H-perfluorononyl carbonate
Formel:
C₁₄H₁₂F₁₆O₃ Molecular Weight: 532,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334453 CAS-Nummer: 1923065-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...