4-Bromo-3,3,4,4-tetrafluorobutyl sec-butyl carbonate
Lieferant: Apollo Scientific
PC450482-25G
APOSPC450482-25GEA
1540
EUR
APOSPC450482-25G
4-Bromo-3,3,4,4-tetrafluorobutyl sec-butyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl sec-butyl carbonate
Formel:
C₉H₁₃BrF₄O₃ Molecular Weight: 325,095 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334475 CAS-Nummer: 1980085-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...