N-Cyclohexyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide 95%
Lieferant: Apollo Scientific
PC450486-1G
APOSPC450486-1GEA
148
EUR
APOSPC450486-1G
N-Cyclohexyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide 95%
N-Cyclohexyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide
Formel:
C₁₀H₁₃F₆NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28901331 CAS-Nummer: 1980087-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...