3-(2,2,3,3-Tetrafluoropropoxy)propyl-1-ene carbonate

Lieferant: Apollo Scientific

PC450494-250MG
APOSPC450494-250MGEA 637 EUR
APOSPC450494-250MG
3-(2,2,3,3-Tetrafluoropropoxy)propyl-1-ene carbonate
3-(2,2,3,3-Tetrafluoropropoxy)propyl-1-ene carbonate
Formel: C₇H₈F₄O₄
Molecular Weight: 232,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28901315
CAS-Nummer: 879496-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR