(2-Bromo-1-ethoxy-2,2-difluoroethoxy)triethylsilane 98%

Lieferant: Apollo Scientific

PC450503-1G
APOSPC450503-1GEA 1020 EUR
APOSPC450503-1G
(2-Bromo-1-ethoxy-2,2-difluoroethoxy)triethylsilane 98%
(2-Bromo-1-ethoxy-2,2-difluoroethoxy)triethylsilane
Formel: C₁₀H₂₁BrF₂O₂Si
Lagertemperatur: Raumtemperatur
MDL: MFCD28901345
CAS-Nummer: 1922958-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR