(1-Ethoxy-2,2,2-trifluoroethoxy)trimethylsilane

Lieferant: Apollo Scientific

PC450525-1G
APOSPC450525-1GEA 477 EUR
APOSPC450525-1G
(1-Ethoxy-2,2,2-trifluoroethoxy)trimethylsilane
(1-Ethoxy-2,2,2-trifluoroethoxy)trimethylsilane
Formel: C₇H₁₅F₃O₂Si
Siedepunkt: 55…56 °C (60 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD30730308
CAS-Nummer: 141023-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR