1,1,1-Trifluoro-2,2-bis(trimethylsiloxy)ethane 97%

Lieferant: Apollo Scientific

PC450526-1G
APOSPC450526-1GEA 760 EUR
APOSPC450526-1G
1,1,1-Trifluoro-2,2-bis(trimethylsiloxy)ethane 97%
1,1,1-Trifluoro-2,2-bis(trimethylsiloxy)ethane
Formel: C₈H₁₉F₃O₂Si₂
Lagertemperatur: Raumtemperatur
MDL: MFCD30730309
CAS-Nummer: 2149590-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR