4,4-Bis(trifluoromethyl)-5,5,6,6,7,7,7-heptafluoroheptyl acrylate 97%

Lieferant: Apollo Scientific
Achtung

PC450547-1G PC450547-5G
APOSPC450547-1GEA 115 EUR
APOSPC450547-1G APOSPC450547-5G
4,4-Bis(trifluoromethyl)-5,5,6,6,7,7,7-heptafluoroheptyl acrylate 97%
4,4-Bis(trifluoromethyl)-5,5,6,6,7,7,7-heptafluoroheptyl acrylate
Formel: C₁₂H₉F₁₃O₂
Molecular Weight: 432.178 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28334481
CAS-Nummer: 1980085-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR