1,4-Bis(trimethylsilyl)tetrafluorobenzene

Lieferant: Apollo Scientific
Achtung

PC450555-100G PC450555-25G
APOSPC450555-100GEA 725 EUR
APOSPC450555-100G APOSPC450555-25G
1,4-Bis(trimethylsilyl)tetrafluorobenzene
1,4-Bis(trimethylsilyl)tetrafluorobenzene
Formel: C₁₂H₁₈F₄Si₂
Molecular Weight: 294.44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00093109
CAS-Nummer: 16956-91-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR