4-(Fluoromethyl)-1,3-dioxolan-2-one

Lieferant: Apollo Scientific
Achtung

PC450562-1G PC450562-250MG
APOSPC450562-1GEA 690 EUR
APOSPC450562-1G APOSPC450562-250MG
4-(Fluoromethyl)-1,3-dioxolan-2-one
4-(Fluoromethyl)-1,3-dioxolan-2-one
Formel: C₄H₅FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22577168
CAS-Nummer: 127213-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR