6-Amino-3-chloro-2-fluorobenzoic acid 96%

Lieferant: Apollo Scientific
Gefahr

PC450566-100MG
APOSPC450566-100MGEA 81.2 EUR
APOSPC450566-100MG
6-Amino-3-chloro-2-fluorobenzoic acid 96%
6-Amino-3-chloro-2-fluorobenzoic acid
Formel: C₇H₅ClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03618452
CAS-Nummer: 874782-54-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR