(Z)-(1,2-Difluorovinyl)trimethylsilane 97%

Lieferant: Apollo Scientific
Gefahr

PC450572-1G PC450572-250MG
APOSPC450572-1GEA 857 EUR
APOSPC450572-1G APOSPC450572-250MG
(Z)-(1,2-Difluorovinyl)trimethylsilane 97%
(Z)-(1,2-Difluorovinyl)trimethylsilane
Formel: C₅H₁₀F₂Si
Lagertemperatur: Raumtemperatur
MDL: MFCD30062987
CAS-Nummer: 17757-16-3
UN: 1993
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR