4-Chlorophenyl-[3-(trifluoromethyl)phenoxy]acetic acid
Lieferant: Apollo Scientific
|
Gefahr
|
PC450575-250MG
APOSPC450575-250MGEA
185
EUR
APOSPC450575-250MG
4-Chlorophenyl-[3-(trifluoromethyl)phenoxy]acetic acid
4-Chlorophenyl-[3-(trifluoromethyl)phenoxy]acetic acid
Formel:
C₁₅H₁₀ClF₃O₃ Molecular Weight: 330,686 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09752899 CAS-Nummer: 4687-08-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...