1H,1H,13H-Tetracosafluor-1-tridecanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC4520-1G
PC4520-25G
PC4520-5G
APOSPC4520-1GEA
23.2
EUR
APOSPC4520-1G
APOSPC4520-25G
APOSPC4520-5G
1H,1H,13H-Tetracosafluor-1-tridecanol 95%
1H,1H,13H-Tetracosafluor-1-tridecanol
Formel:
C₁₃H₄OF₂₄ Schmelzpunkt: 129…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155920 CAS-Nummer: 423-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...