1H,1H,9H-Hexadecafluor-1-nonanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4530-100G
PC4530-250G
PC4530-25G
APOSPC4530-100GEA
92.8
EUR
APOSPC4530-100G
APOSPC4530-250G
APOSPC4530-25G
1H,1H,9H-Hexadecafluor-1-nonanol 97%
1H,1H,9H-Hexadecafluor-1-nonanol
Contains upto 10% H(CF2CF2)nCHOH(CF2CF2)mH where m+n=4.
Formel:
C₉H₄F₁₆O Siedepunkt: 155…158 °C/200mmHg Schmelzpunkt: 53…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039629 CAS-Nummer: 376-18-1 EINECS: 206-806-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...