6-(Trifluoromethyl)thionicotinamide 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC4545-250MG
APOSPC4545-250MGEA
21.9
EUR
APOSPC4545-250MG
6-(Trifluoromethyl)thionicotinamide 95+%
6-(Trifluoromethyl)thionicotinamide
Formel:
C₇H₅F₃N₂S Schmelzpunkt: 160…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02173905 CAS-Nummer: 386715-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...