2-Bromo-1-chloro-4-trifluoromethoxybenzene 98%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-1-chloro-4-(trifluoromethoxy)benzene

PC4547-100G
APOSPC4547-100GEA 139 EUR
APOSPC4547-100G
2-Bromo-1-chloro-4-trifluoromethoxybenzene 98%
2-Bromo-1-chloro-4-trifluoromethoxybenzene
Formel: C₇H₃BrClF₃O
Molecular Weight: 275,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797751
CAS-Nummer: 468075-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR