2-Methyl-6-(trifluoromethyl)nicotinonitrile

Lieferant: Apollo Scientific

PC4552-1G
APOSPC4552-1GEA 145 EUR
APOSPC4552-1G
2-Methyl-6-(trifluoromethyl)nicotinonitrile
2-Methyl-6-(trifluoromethyl)nicotinonitrile
Formel: C₈H₅F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01311987
CAS-Nummer: 402479-93-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR