3-(Trifluoroacetyl)coumarin

Lieferant: Apollo Scientific
Achtung

PC4569-1G
APOSPC4569-1GEA 174 EUR
APOSPC4569-1G
3-(Trifluoroacetyl)coumarin
3-(Trifluoroacetyl)coumarin
Formel: C₁₁H₅F₃O₃
Molecular Weight: 242,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094246
CAS-Nummer: 503315-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR