2-Amino-3'-fluoracetophenon Hydrochlorid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluorophenacylamine hydrochloride

PC4574-250MG
APOSPC4574-250MGEA 21.9 EUR
APOSPC4574-250MG
2-Amino-3'-fluoracetophenon Hydrochlorid 98%
2-Amino-3'-fluoracetophenon Hydrochlorid
Formel: C₈H₉ClFNO
Schmelzpunkt: 201…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08532459
CAS-Nummer: 93102-97-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR