1,1,1,2,3,3-Hexafluor-2-(heptafluorpropoxy)-3-[(trifluorvinyl)oxy]propan
Lieferant: Apollo Scientific
|
Achtung
|
PC4581-1G
PC4581-25G
PC4581-5G
APOSPC4581-1GEA
33.5
EUR
APOSPC4581-1G
APOSPC4581-25G
APOSPC4581-5G
1,1,1,2,3,3-Hexafluor-2-(heptafluorpropoxy)-3-[(trifluorvinyl)oxy]propan
1,1,1,2,3,3-Hexafluor-2-(heptafluorpropoxy)-3-[(trifluorvinyl)oxy]propan
Formel:
F₂CCFOCF₂CF(CF₃)O(CF₂)₂CF₃ Siedepunkt: 103…104 °C Dichte: 1,8 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00191479 CAS-Nummer: 1644-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...