2,5-Difluorphenylhydrazin 98%

Lieferant: Apollo Scientific
Achtung

PC4585-1G
APOSPC4585-1GEA 21.9 EUR
APOSPC4585-1G
2,5-Difluorphenylhydrazin 98%
2,5-Difluorphenylhydrazin
Formel: C₆H₆F₂N₂
Molecular Weight: 144,12 g/mol
Schmelzpunkt: 73…76 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00013384
CAS-Nummer: 97108-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR