2-Fluor-5-methylphenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-5-methylbenzeneboronic acid, 2-Fluoro-5-methylboronic acid
PC4606-25G
PC4606-5G
PC4606-100G
APOSPC4606-25GEA
59.6
EUR
APOSPC4606-25G
APOSPC4606-5G
APOSPC4606-100G
2-Fluor-5-methylphenylboronsäure 95%
2-Fluor-5-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153.9466 g/mol Schmelzpunkt: 77…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03428512 CAS-Nummer: 166328-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...