2-Fluor-5-methylphenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-methylbenzeneboronic acid, 2-Fluoro-5-methylboronic acid

PC4606-25G PC4606-5G PC4606-100G
APOSPC4606-25GEA 59.6 EUR
APOSPC4606-25G APOSPC4606-5G APOSPC4606-100G
2-Fluor-5-methylphenylboronsäure 95%
2-Fluor-5-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153.9466 g/mol
Schmelzpunkt: 77…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03428512
CAS-Nummer: 166328-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR