3-Chlor-5-fluoranilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4608-5G
PC4608-1G
PC4608-25G
APOSPC4608-5GEA
34.8
EUR
APOSPC4608-5G
APOSPC4608-1G
APOSPC4608-25G
3-Chlor-5-fluoranilin 98%
3-Chlor-5-fluoranilin
Formel:
C₆H₅ClFN Dichte: 1,45 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD03407962 CAS-Nummer: 4863-91-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...