3-Carbamoyl-4-fluorobenzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Aminocarbonyl)-4-fluorophenylboronic acid, (3-Carbamoyl-4-fluorophenyl)boronic acid

PC4611-1G
APOSPC4611-1GEA 261 EUR
APOSPC4611-1G
3-Carbamoyl-4-fluorobenzeneboronic acid 97%
3-Carbamoyl-4-fluorobenzeneboronic acid
Formel: C₇H₇BFNO₃
Molecular Weight: 182,95 g/mol
Schmelzpunkt: 257…259 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235053
CAS-Nummer: 874219-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR