3-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-5-(morpholine-4-carbonyl)phenylboronic acid, 3-Fluoro-5-(4-morpholinylcarbonyl)benzeneboronic acid

PC4619-1G
APOSPC4619-1GEA 205 EUR
APOSPC4619-1G
3-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure 98%
3-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₄
Molecular Weight: 253,04 g/mol
Schmelzpunkt: 145…147 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235057
CAS-Nummer: 874219-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR