1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: O-(4-Fluorobenzyl)hydroxylamine hydrochloride

PC4627-1G
APOSPC4627-1GEA 21.9 EUR
APOSPC4627-1G
1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid
1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid
Formel: C₇H₉ClFNO
Molecular Weight: 177,6039 g/mol
Schmelzpunkt: 225…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01114581
CAS-Nummer: 51572-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR